missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butoxy-2,3-difluorobenzene, 95%, Thermo Scientific™

Catalog Number p-7054048
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
136239-66-2
C10H12F2O
186.202
MFCD07368738
HYDIPEHDSUNHPA-UHFFFAOYSA-N
3-butoxy-1,2-difluorobenzene, 2,3-difluorobutoxybenzene, benzene, 1-butoxy-2,3-difluoro, 1-butoxy-2,3-difluoro-benzene, pubchem15526, acmc-209c5k, ksc493i5l, 2,3-difluoro-1-butoxybenzene, 4-butoxy-2,3-difluorobenzene
2782939
1-butoxy-2,3-difluorobenzene
CCCCOC1=C(C(=CC=C1)F)F
This item is not returnable. View return policy
136239-66-2
C10H12F2O
186.202
MFCD07368738
HYDIPEHDSUNHPA-UHFFFAOYSA-N
3-butoxy-1,2-difluorobenzene, 2,3-difluorobutoxybenzene, benzene, 1-butoxy-2,3-difluoro, 1-butoxy-2,3-difluoro-benzene, pubchem15526, acmc-209c5k, ksc493i5l, 2,3-difluoro-1-butoxybenzene, 4-butoxy-2,3-difluorobenzene
2782939
1-butoxy-2,3-difluorobenzene
CCCCOC1=C(C(=CC=C1)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 5 g
Formula Weight 186.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-n-Butoxy-2,3-difluorobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'