missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-3779131
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19012-03-4
C10H9NO
159.19
MFCD00014570
KXYBYRKRRGSZCX-UHFFFAOYSA-N
1-methyl-1h-indole-3-carbaldehyde, 1-methylindole-3-carboxaldehyde, 1-methyl-indole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methylindole, 1-methyl-3-formylindole, n-methyl-3-formylindole, indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methyl-1h-indole, n-methyl-3-indolecarboxaldehyde
87894
1-methylindole-3-carbaldehyde
CN1C=C(C=O)C2=CC=CC=C12
This item is not returnable. View return policy
19012-03-4
C10H9NO
159.19
MFCD00014570
KXYBYRKRRGSZCX-UHFFFAOYSA-N
1-methyl-1h-indole-3-carbaldehyde, 1-methylindole-3-carboxaldehyde, 1-methyl-indole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methylindole, 1-methyl-3-formylindole, n-methyl-3-formylindole, indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methyl-1h-indole, n-methyl-3-indolecarboxaldehyde
87894
1-methylindole-3-carbaldehyde
CN1C=C(C=O)C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 72°C
Color Brown or Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 159.19
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-Methylindole-3-carboxaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'