Learn More
1-Methyl-3-n-octylimidazolium chloride, 97%, Thermo Scientific Chemicals
Catalog Number p-7053281
Click to view available options
Quantity:
5 g
50 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
64697-40-1
C12H23ClN2
230.78
MFCD03095432
OXFBEEDAZHXDHB-UHFFFAOYSA-M
1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j
2734223
CHEBI:61342
1-methyl-3-octylimidazol-1-ium;chloride
[Cl-].CCCCCCCCN1C=C[N+](C)=C1
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
64697-40-1
C12H23ClN2
230.78
MFCD03095432
OXFBEEDAZHXDHB-UHFFFAOYSA-M
1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j
2734223
CHEBI:61342
1-methyl-3-octylimidazol-1-ium;chloride
[Cl-].CCCCCCCCN1C=C[N+](C)=C1
Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.01 g/mL
Refractive Index 1.51
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 230.78
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium chloride