missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium thiocyanate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
Description
It is used as a pharmaceutical intermediate and also used as primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate and also used as primary and secondary intermediate.
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
It is used as a pharmaceutical intermediate and also used as primary and secondary intermediate.
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Specifications
Melting Point | -6°C |
Density | 1.14 |
Refractive Index | n20/D 1.553 |
Quantity | 5 g |
UN Number | UN2810 |
Sensitivity | Moisture sensitive |
Formula Weight | 169.25 |
Percent Purity | 98% |
Chemical Name or Material | 1-Ethyl-3-methylimidazolium thiocyanate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'