missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals

Catalog Number p-3778497
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2386-53-0
MFCD00007527
DAJSVUQLFFJUSX-UHFFFAOYSA-M
sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
23665726
sodium;dodecane-1-sulfonate
CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
2386-53-0
MFCD00007527
DAJSVUQLFFJUSX-UHFFFAOYSA-M
sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
23665726
sodium;dodecane-1-sulfonate
CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY
Absorbance 0.02 max. (C=0.18 M in hot water) at 230nm, 0.03 max. (C=0.18 M in hot water) at 220nm, 0.05 max. (C=0.18 M in hot water) at 210nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point >300.0°C
Color White
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C12H25NaO3S
Linear Formula CH3(CH2)11SO3Na
Quantity 5 g
Solubility Information Solubility in water: soluble.
Molecular Weight (g/mol) 272.38
Formula Weight 272.38
Percent Purity 99+%
Grade Ion Pair Chromatography
Physical Form Crystalline Powder
Chemical Name or Material 1-Dodecanesulfonic acid, sodium salt, Anhydrous, Ion pair chromatography
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'