missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-heptyloxybenzene, 97%, Thermo Scientific™

Catalog Number 11466626
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
123732-04-7
C13H19BrO
271.198
MFCD00173752
DSCVEUSJTAFLBF-UHFFFAOYSA-N
1-4-bromophenoxy heptane, 1-bromo-4-heptyloxybenzene, 1-bromo-4-heptyloxy benzene, 4-n-heptyloxybromobenzene, 1-bromo-4-n-heptyloxybenzene, 4-bromophenylheptyl ether, 4-bromo-heptyloxy benzene
599102
1-bromo-4-heptoxybenzene
CCCCCCCOC1=CC=C(C=C1)Br
This item is not returnable. View return policy
123732-04-7
C13H19BrO
271.198
MFCD00173752
DSCVEUSJTAFLBF-UHFFFAOYSA-N
1-4-bromophenoxy heptane, 1-bromo-4-heptyloxybenzene, 1-bromo-4-heptyloxy benzene, 4-n-heptyloxybromobenzene, 1-bromo-4-n-heptyloxybenzene, 4-bromophenylheptyl ether, 4-bromo-heptyloxy benzene
599102
1-bromo-4-heptoxybenzene
CCCCCCCOC1=CC=C(C=C1)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 271.2
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-n-heptyloxybenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'