missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-methylimidazolium tetrafluoroborate, 97%, Thermo Scientific™

Catalog Number p-7053364
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
500996-04-3
C11H13BF4N2
260.04
MFCD07784449
QULDEUUWXMCUFO-UHFFFAOYSA-N
1-benzyl-3-methylimidazolium tetrafluoroborate, 1-benzyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate, 3-benzyl-1-methylimidazol-1-ium tetrafluoroborate, 1-benzyl-3-methylimidazol-1-ium tetrafluoroborate, 1-benzyl-3-methylimidazolium tetrafluoroborate hplc
12138923
F[B-](F)(F)F.C[N+]1=CN(CC2=CC=CC=C2)C=C1
This item is not returnable. View return policy
500996-04-3
C11H13BF4N2
260.04
MFCD07784449
QULDEUUWXMCUFO-UHFFFAOYSA-N
1-benzyl-3-methylimidazolium tetrafluoroborate, 1-benzyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate, 3-benzyl-1-methylimidazol-1-ium tetrafluoroborate, 1-benzyl-3-methylimidazol-1-ium tetrafluoroborate, 1-benzyl-3-methylimidazolium tetrafluoroborate hplc
12138923
F[B-](F)(F)F.C[N+]1=CN(CC2=CC=CC=C2)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Sensitivity Moisture sensitive
IUPAC Name 1-benzyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Formula Weight 260.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Benzyl-3-methylimidazolium tetrafluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'