missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluoro-2-nitrophenyl)piperidine, 97%

Brand:  Alfa Aesar™ H63142.06

Additional Details : CAS Number : 719-70-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15459668

  • 151.00 EUR / 5g


Estimated Shipment date
from Supplier 21-02-2019
Add to basket

Specifications

Chemical Identifiers

719-70-0
224.235
YSBDMNNCMGUEAL-UHFFFAOYSA-N
2737434
C1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
C11H13FN2O2
MFCD00798370
1-4-fluoro-2-nitrophenyl piperidine, n-4-fluoro-2-nitro-phenyl-piperidine, n-4-fluoro-2-nitro-phenyl piperidine, pubchem8030, acmc-1bbsb, 4-fluoro-2-nitrophenyl piperidine, piperidine,1-4-fluoro-2-nitrophenyl, 1-4-fluoranyl-2-nitro-phenyl piperidine
1-(4-fluoro-2-nitrophenyl)piperidine

Specifications

1-(4-Fluoro-2-nitrophenyl)piperidine
719-70-0
MFCD00798370
YSBDMNNCMGUEAL-UHFFFAOYSA-N
1-(4-fluoro-2-nitrophenyl)piperidine
2737434
Solid
5g
C11H13FN2O2
1-4-fluoro-2-nitrophenyl piperidine, n-4-fluoro-2-nitro-phenyl-piperidine, n-4-fluoro-2-nitro-phenyl piperidine, pubchem8030, acmc-1bbsb, 4-fluoro-2-nitrophenyl piperidine, piperidine,1-4-fluoro-2-nitrophenyl, 1-4-fluoranyl-2-nitro-phenyl piperidine
C1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
224.235
224.23
97%
Videos
SDS