missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluoro-2-nitrophenyl)piperazine, 97%

Brand:  Alfa Aesar™ H63133.03

Additional Details : CAS Number : 243128-46-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459658

  • 75.70 EUR / 1g


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

243128-46-3
225.223
ZPYLKZRKXDCEQI-UHFFFAOYSA-N
2737435
C10H12FN3O2
MFCD00798371
1-4-fluoro-2-nitrophenyl piperazine, 4-fluoro-2-nitrophenyl piperazine, piperazine, 1-4-fluoro-2-nitrophenyl, 1-4-fluoro-2-nitro-phenyl piperazine, 5-fluoro-2-piperazin-1-yl nitrobenzene
C1CN(CCN1)C2=C(C=C(C=C2)F)[N+](=O)[O-]

Specifications

1-(4-Fluoro-2-nitrophenyl)piperazine
243128-46-3
MFCD00798371
ZPYLKZRKXDCEQI-UHFFFAOYSA-N
225.223
225.22
97%
1g
C10H12FN3O2
1-4-fluoro-2-nitrophenyl piperazine, 4-fluoro-2-nitrophenyl piperazine, piperazine, 1-4-fluoro-2-nitrophenyl, 1-4-fluoro-2-nitro-phenyl piperazine, 5-fluoro-2-piperazin-1-yl nitrobenzene
C1CN(CCN1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
2737435
Solid
Videos
SDS