missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Thermo Scientific Chemicals

Catalog Number p-7056468
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
369-77-7
C14H9Cl2F3N2O
349.13
MFCD00867294
ZFSXZJXLKAJIGS-UHFFFAOYSA-N
cloflucarban, halocarban, irgasan cf3, cloflucarbon, trifluoromethyldichlorocarbanilide, unii-i5zzy3dc5g, cloflucarban usan, 4,4'-dichloro-3-trifluoromethyl carbanilide, i5zzy3dc5g, halocarban inn
9719
1-(4-chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
FC(F)(F)C1=CC(NC(=O)NC2=CC=C(Cl)C=C2)=CC=C1Cl
This item is not returnable. View return policy
369-77-7
C14H9Cl2F3N2O
349.13
MFCD00867294
ZFSXZJXLKAJIGS-UHFFFAOYSA-N
cloflucarban, halocarban, irgasan cf3, cloflucarbon, trifluoromethyldichlorocarbanilide, unii-i5zzy3dc5g, cloflucarban usan, 4,4'-dichloro-3-trifluoromethyl carbanilide, i5zzy3dc5g, halocarban inn
9719
1-(4-chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
FC(F)(F)C1=CC(NC(=O)NC2=CC=C(Cl)C=C2)=CC=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 349.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'