Learn More
Alfa Aesar™ 1-(4-Bromo-3-methylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%
Brand:  Alfa Aesar™ H65105.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15459376
  • 270.00 EUR / 1g
Estimated Shipment date
from Supplier 11-06-2018
Add to basket
Specifications
Chemical Identifiers
1-(4-Bromo-3-methylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
407.62
MFCD27975719
Specifications
1-(4-Bromo-3-methylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
1g
407.62
Solid
97%
MFCD27975719