missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanedisulfonic acid, 70% w/v aq. soln., Thermo Scientific™

Catalog Number 11470407
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
21668-77-9
C3H6O6S2
202.20
MFCD00191482
MGNVWUDMMXZUDI-UHFFFAOYSA-L
1,3-propanedisulfonic acid, eprodisate, 1,3-propanedisulfonic acid in water, unii-6qfp76v7s7, propane-1,3-disulfonate, eprodisate inn, 1,3-propanesulfonic acid, 1,3-propanedisulfonicacid, 1,3-propanedisulphonic acid, 1,3 propanesulfonate
428573
propane-1,3-disulfonic acid
[O-]S(=O)(=O)CCCS([O-])(=O)=O
This item is not returnable. View return policy
21668-77-9
C3H6O6S2
202.20
MFCD00191482
MGNVWUDMMXZUDI-UHFFFAOYSA-L
1,3-propanedisulfonic acid, eprodisate, 1,3-propanedisulfonic acid in water, unii-6qfp76v7s7, propane-1,3-disulfonate, eprodisate inn, 1,3-propanesulfonic acid, 1,3-propanedisulfonicacid, 1,3-propanedisulphonic acid, 1,3 propanesulfonate
428573
propane-1,3-disulfonic acid
[O-]S(=O)(=O)CCCS([O-])(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.33
Refractive Index 1.422
Quantity 25 g
UN Number UN3265
Formula Weight 204.21
Concentration or Composition (by Analyte or Components) 70% w/v aq. soln.
Chemical Name or Material 1,3-Propanedisulfonic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'