missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyluracil-5-carboxaldehyde, 96%, Thermo Scientific™

Catalog Number 11419430
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
4869-46-9
C7H8N2O3
168.15
MFCD00099582
KGJFRZOATIXYPW-UHFFFAOYSA-N
1,3-dimethyluracil-5-carboxaldehyde, 1,3-dimethyl-5-formyluracil, 1,3-dime-5-formu, 1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde, 1,2,3,4-tetrahydro-1,3-dimethyl-2,4-dioxo, 1,3-dimethyl-2,6-dioxo-1,3-dihydropyrimidine-5-carbaldehyde, acmc-1asas, 5-formyl-1,3-dimethyluracil, 1,3-dimethyl-2,4-dioxo-pyrimidine-5-carbaldehyde, 1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin
453092
1,3-dimethyl-2,4-dioxopyrimidine-5-carbaldehyde
CN1C=C(C=O)C(=O)N(C)C1=O
This item is not returnable. View return policy
4869-46-9
C7H8N2O3
168.15
MFCD00099582
KGJFRZOATIXYPW-UHFFFAOYSA-N
1,3-dimethyluracil-5-carboxaldehyde, 1,3-dimethyl-5-formyluracil, 1,3-dime-5-formu, 1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde, 1,2,3,4-tetrahydro-1,3-dimethyl-2,4-dioxo, 1,3-dimethyl-2,6-dioxo-1,3-dihydropyrimidine-5-carbaldehyde, acmc-1asas, 5-formyl-1,3-dimethyluracil, 1,3-dimethyl-2,4-dioxo-pyrimidine-5-carbaldehyde, 1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin
453092
1,3-dimethyl-2,4-dioxopyrimidine-5-carbaldehyde
CN1C=C(C=O)C(=O)N(C)C1=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 168.15
Percent Purity 96%
Chemical Name or Material 1,3-Dimethyluracil-5-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'