missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Chloro-4-fluorophenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62106.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447036

  • 323.00 EUR / 1g


Estimated Shipment date
from Supplier 18-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD00430128

Specifications

1-(3-Chloro-4-fluorophenyl)-3-(4-fluorophenyl)urea
MFCD00430128
Solid
1g
282.67
97%
Videos
SDS