missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluoro-4-nitrophenyl)piperazine, 97%, Thermo Scientific™

Catalog Number 15459528
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
154590-33-7
C10H12FN3O2
225.223
MFCD01572463
ZILOTWJFFLIFMZ-UHFFFAOYSA-N
2783066
C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])F
This item is not returnable. View return policy
154590-33-7
C10H12FN3O2
225.223
MFCD01572463
ZILOTWJFFLIFMZ-UHFFFAOYSA-N
2783066
C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Synonym 1-2-fluoro-4-nitrophenyl piperazine, piperazine, 1-2-fluoro-4-nitrophenyl, 1-2-fluoro-4-nitro-phenyl-piperazine, 2-fluoro-4-nitrophenyl piperazine, piperazine,1-2-fluoro-4-nitrophenyl, buttpark 153\33-48, 1-2-fluoranyl-4-nitro-phenyl piperazine
Formula Weight 225.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'