missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloropyridin-4-yl)ethanone, 97%, Thermo Scientific™

Catalog Number p-3522394
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
23794-15-2
C7H6ClNO
155.581
MFCD07699406
ZJCGPQZERFBGSM-UHFFFAOYSA-N
1-2-chloropyridin-4-yl ethanone, 2-chloro-4-acetylpyridine, 4-acetyl-2-chloropyridine, 1-2-chloropyridine-4-yl ethanone, 1-2-chloropyridin-4-yl ethan-1-one, 1-2-chloro-pyridin-4-yl-ethanone, 1-2-chloro-4-pyridinyl-1-ethanone, ethanone, 1-2-chloro-4-pyridinyl, 1-2-chloro-4-pyridyl ethanone, 1-2-chloro-4-pyridinyl ethanone
13145255
1-(2-chloropyridin-4-yl)ethanone
CC(=O)C1=CC(=NC=C1)Cl
This item is not returnable. View return policy
23794-15-2
C7H6ClNO
155.581
MFCD07699406
ZJCGPQZERFBGSM-UHFFFAOYSA-N
1-2-chloropyridin-4-yl ethanone, 2-chloro-4-acetylpyridine, 4-acetyl-2-chloropyridine, 1-2-chloropyridine-4-yl ethanone, 1-2-chloropyridin-4-yl ethan-1-one, 1-2-chloro-pyridin-4-yl-ethanone, 1-2-chloro-4-pyridinyl-1-ethanone, ethanone, 1-2-chloro-4-pyridinyl, 1-2-chloro-4-pyridyl ethanone, 1-2-chloro-4-pyridinyl ethanone
13145255
1-(2-chloropyridin-4-yl)ethanone
CC(=O)C1=CC(=NC=C1)Cl
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 155.58
Percent Purity 97%
Chemical Name or Material 1-(2-Chloropyridin-4-yl)ethanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'