missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Alfa Aesar™

Brand:  Alfa Aesar H65119.MD

Additional Details : CAS Number : 418805-03-5 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15459386

  • 84.00 EUR / 250mg


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

418805-03-5
MFCD01654414
1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-chlorophenyl)urea

Specifications

1-(2-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
418805-03-5
1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-chlorophenyl)urea
Solid
250mg
MFCD01654414
349.13
97%
Videos
SDS