missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(diphenylphosphino)benzene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
Description
It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.
Solubility
Insoluble in water.
Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.
Solubility
Insoluble in water.
Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Melting Point | 185°C to 189°C |
Quantity | 0.25 g |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 446.47 |
Percent Purity | 98% |
Chemical Name or Material | 1,2-Bis(diphenylphosphino)benzene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'