missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,4-Difluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61329.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486436

  • 646.00 EUR / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,4-Difluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
316.23
MFCD06062278

Specifications

1-(2,4-Difluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
1g
97%
MFCD06062278
White
Solid
316.23
Videos
SDS