Learn More
Alfa Aesar™ 1-(2,4-Difluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%
Brand:  Alfa Aesar™ H64622.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15459026
  • 1345.00 EUR / 5g
Estimated Shipment date
from Supplier 11-06-2018
Add to basket
Specifications
Chemical Identifiers
1-(2,4-Difluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
332.23
MFCD05990291
Specifications
1-(2,4-Difluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
332.23
5g
97%
MFCD05990291