Learn More
1,2,3-Benzothiadiazole-5-carbonyl chloride, 95%, Thermo Scientific™
Catalog Number p-4509998
Click to view available options
Quantity:
250 mg
1 g
5 g
321309-32-4
C7H3ClN2OS
198.624
VOSTUOLSYXVSND-UHFFFAOYSA-N
1,2,3-benzothiadiazole-5-carbonylchloride, benzo d 1,2,3-thiadiazole-5-carbonyl chloride, benzo d 1,2,3 thiadiazole-5-carbonyl chloride
2735457
1,2,3-benzothiadiazole-5-carbonyl chloride
C1=CC2=C(C=C1C(=O)Cl)N=NS2
321309-32-4
C7H3ClN2OS
198.624
VOSTUOLSYXVSND-UHFFFAOYSA-N
1,2,3-benzothiadiazole-5-carbonylchloride, benzo d 1,2,3-thiadiazole-5-carbonyl chloride, benzo d 1,2,3 thiadiazole-5-carbonyl chloride
2735457
1,2,3-benzothiadiazole-5-carbonyl chloride
C1=CC2=C(C=C1C(=O)Cl)N=NS2
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 198.63
Percent Purity 95%
Chemical Name or Material 1, 2, 3-Benzothiadiazole-5-carbonyl chloride