missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Bis(dichlorophosphino)ferrocene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
It is applied as a reactant for synthesis of ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation, metallocene-bridged bisphosphaalkene and as a catalyst for synthesis of oligonuclear heterometallic complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as a reactant for synthesis of ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation, metallocene-bridged bisphosphaalkene and as a catalyst for synthesis of oligonuclear heterometallic complexes.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is applied as a reactant for synthesis of ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation, metallocene-bridged bisphosphaalkene and as a catalyst for synthesis of oligonuclear heterometallic complexes.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Quantity | 1 g |
UN Number | UN3261 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 387.78 |
Percent Purity | 98% |
Chemical Name or Material | 1,1'-Bis(dichlorophosphino)ferrocene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'