missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,4-Tetraphenyl-1,3-butadiene, 99%, Thermo Scientific Chemicals

Catalog Number p-198705
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1450-63-1
C28H22
358.48
MFCD00004766
KLCLIOISYBHYDZ-UHFFFAOYSA-N
1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene
74060
1,4,4-triphenylbuta-1,3-dienylbenzene
C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1450-63-1
C28H22
358.48
MFCD00004766
KLCLIOISYBHYDZ-UHFFFAOYSA-N
1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene
74060
1,4,4-triphenylbuta-1,3-dienylbenzene
C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 196.0°C to 200.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)2C=CHCH=C(C6H5)2
Quantity 1 g
Beilstein 05, 750
Formula Weight 358.47
Percent Purity 99%
Physical Form Needle-like Crystals
Chemical Name or Material 1,1,4,4-Tetraphenyl-1,3-butadiene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'