man

Test

CAS RN 5336-75-4

H3CNCH3N

CAS RN 5336-75-4

IUPAC Name: 4-(diethylamino)butanenitrile
Synonyms: 4-(diethylamino)butanenitrile
Molecular Weight (g/mol): 140.23
Molecular Formula: C8H16N2
InChi Key: NUVVGLXJJPTXRJ-UHFFFAOYSA-N
SMILES: CCN(CC)CCCC#N
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results