accessibility menu, dialog, popup

UserName

CAS RN 137348-86-8

CH3CH3NH3CCH3POCH3H3CCH3OCH3H3CCH3

CAS RN 137348-86-8

IUPAC Name: [bis(tert-butoxy)phosphanyl]bis(propan-2-yl)amine
Synonyms: [bis(tert-butoxy)phosphanyl]diisopropylamine
Molecular Weight (g/mol): 277.39
Molecular Formula: C14H32NO2P
InChi Key: YGFLCNPXEPDANQ-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)P(OC(C)(C)C)OC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)