accessibility menu, dialog, popup

Test

CAS RN 106946-74-1

CAS RN 106946-74-1

IUPAC Name: tert-butyl N-[(2S)-1-hydroxy-3-methylpentan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-hydroxy-3-methylpentan-2-yl]carbamate
Molecular Weight (g/mol): 217.31
Molecular Formula: C11H23NO3
InChi Key: BPLDQMXXYMKQPW-YGPZHTELSA-N
SMILES: CCC(C)[C@@H](CO)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)