CAS RN 1061605-21-7
CAS RN 1061605-21-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Thalidomide-4-hydroxyacetate
Carboxylic acid-functionalized Thalidomide; cereblon ligand for PROTACs
2
Thalidomide-O-COOH, MedChemExpress
MedChemExpress Thalidomide-O-COOH (Cereblon ligand 3) is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide-O-COOH (Cereblon ligand 3) can be connected to the ligand for protein by a linker to form PROTACs.

3
TC E3 5031, Tocris Bioscience™
Carboxylic acid-functionalized Thalidomide; cereblon ligand for PROTACs
