CAS RN 105875-75-0
CAS RN 105875-75-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Allyloxy-tert-butyldimethylsilane, 98%, Thermo Scientific Chemicals
CAS: 105875-75-0 Molecular Formula: C9H20OSi MDL Number: MFCD00239355