Fisher Scientific toivottaa tervetulleeksi
Fermentas Molecular Biology Products - Now Available through Fisher Scientific
For over 30 years, Fermentas has been a world leader in the discovery and manufacturing of quality molecular biology products in the fields of genomics, proteomics and microfluidics. All Fermentas products are manufactured in class D clean room facilities, qualified and certified as per EU directives and ISPE guidelines, which are pre-requisites for GMP manufacturing. Fermentas quality assurance is carried out according to ISO9001, ISO13485 quality and ISO14001 environmental management systems, guaranteeing batch-to-batch reproducibility.
2010-11 Catalogue Cover - Click here to download as a PDF Download a copy of the 2010-11 Fermentas product catalogue [PDF - 30MB] Search for Fermentas products
Nucleic acid purification
The GeneJET™ nucleic acid purification range offers a simple, rapid and cost-effective system for the isolation of high quality RNA and DNA from a variety of different sources.
Nucleic Acid Purification Brochure [PDF - 825KB]
cDNA synthesis
Thermo Scientific Fermentas is a specialist in engineering new advanced reverse transcription enzymes. Maxima® Reverse Transcriptase and RevertAid™ Premium Reverse Transcriptase are derived by in vitro evolution of M-MuLV RT. These new enzymes feature highest thermostability and speed among M-MuLV RT derivatives available in the market.
PCR, qPCR, RT-PCR, & dNTPs Brochure [PDF - 1.35MB]
PCR products
These include the DreamTaq enzymes and PCR master mixes for all common PCR applications: routine, hot start, high fidelity and long PCR.
PCR, qPCR, RT-PCR, & dNTPs Brochure [PDF - 1.35MB]
Nucleotides
These are aqueous solutions of deoxynucleotides dATP, dCTP, dGTP and dTTP of >99% purity. Nucleotides are available as solutions of individual dNTPs and as dNTP mixes.
Nucleotides & Primers Brochure [PDF - 663KB]
Primers
Primers are available for cDNA synthesis, PCR and sequencing.
Nucleotides & Primers Brochure [PDF - 663KB]
qPCR products
Maxima® qPCR Master Mixes are a line of optimized products for real time PCR using SYBR Green or probe-based detection. Five different mixes are optimized for use with all types of real time qPCR instruments.
PCR, qPCR, RT-PCR, & dNTPs Brochure [PDF - 1.35MB]
Restriction enzymes
Thermo Scientific Fermentas offers two lines of restriction enzymes:
  • FastDigest® restriction enzymes are an advanced line of restriction enzymes for rapid DNA digestion. All FastDigest® enzymes are 100% active in the universal FastDigest® Green buffer and are able to digest DNA in 5-15 min.
  • Conventional Restriction Enzymes: The product range consists of 201 different enzymes. Every enzyme is provided with a buffer that is optimized for maximal activity, and also a dedicated Tango™ buffer for double digestion.
FastDigest® & Conventional Restriction Enzymes Brochure [PDF - 5.7MB]
Modifying enzymes & in vitro transcription
This consists of a range of high quality DNA/RNA modifying enzymes such as polymerases, ligases, alkaline phosphatases, kinases and nucleases.
In vitro Transcription Brochure [PDF - 555KB] DNA/RNA Modifying Enzymes Brochure [PDF - 1.1MB]
Molecular cloning
Thermo Scientific Fermentas offers enzymes and kits for efficient, fast and simple cloning of DNA with the CloneJET™ positive selection system for PCR product, cloning as an example of highest speed and simplicity.
Molecular Cloning Brochure [PDF - 804KB]
Molecular labelling
Kits and enzymes for the generation of labelled DNA probes.
Molecular Labeling & Detection Brochure [PDF - 571KB]
Electrophoresis reagents
Loading buffers, running buffers and agarose for DNA and RNA agarose gel electrophoresis.
DNA Electrophoresis Brochure [PDF - 1.1MB] RNA Electrophoresis Brochure [PDF - 475KB]
Molecular weight ladders
A wide range of DNA and RNA ladders are available, including the popular GeneRuler™ 1kb and 100 bp ladders. All our DNA ladders are consist of HPLC purified individual DNA fragments and form exceptionally clear sharp bands on gels.
DNA Electrophoresis Brochure [PDF - 1.1MB] RNA Electrophoresis Brochure [PDF - 475KB]
Transfection
TurboFect™ transfection reagents are based on a novel proprietary cationic polymers and provide easy and efficient tools to transfect DNA, siRNA and proteins.
Transfection Brochure [PDF - 590KB]