Learn More
Fisherbrand™ UV Lamp, Dual Wavelength
Used as a method of purification. Fisher Scientific™ UV Lamp, Dual Wavelength can be used in combination with other methods to reduce microbial reproduction in purified water.
Brand:  Fisherbrand™
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15449689
  • 354.00 EUR / Each
Estimated Shipment: 08-10-2024
Description
  • For use with Accu100 and Accu500 Ultrapure Water Systems
  • Prevents microbial reproduction
Specifications
Specifications
185/254nm
Accu100 Ultrapure Water System