Learn More
Haldenwanger™ Unglazed Porcelain Pestle
Use these pestles for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Pestles are constructed from optimized laboratory porcelain.
15.80 EUR - 30.55 EUR
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12313338
|
Haldenwanger™
56/0A |
125mm |
15.80 EUR
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10656405
|
Haldenwanger™
L 56/00/GLASIERT |
115mm |
27.70 EUR
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
10287274
|
Haldenwanger™
L 56/1/RAUH |
135mm |
17.05 EUR
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
10042964
|
Haldenwanger™
L 56/2/RAUH |
150mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12343338
|
Haldenwanger™
L 56/3/RAUH |
175mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Available in a variety of sizes
- Grinding surfaces are unglazed
- Complies with DIN VDE 0335, type C 100
Specifications
Unglazed | |
Pestle | |
Porcelain |