Learn More
DuPont™ Tyvek™ IsoClean™ Slip-retardant Shoe Cover
113.02 EUR valid until 2024-12-31
Use promo code "22918" to get your promotional price.
115.92 EUR valid until 2024-05-31
Use promo code "21724" to get your promotional price.
Use promo code "22918" to get your promotional price.
115.92 EUR valid until 2024-05-31
Use promo code "21724" to get your promotional price.
Click to view available options
Size:
Medium
Large
Description
- Gripper™ soles provide enhanced slip-retardance and robustness
- Delivers an ideal balance of protection, durability and comfort li>Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms
Specifications
Size | Medium |
Color | White |
Sterility | Non-sterile |
Model | IC451S00 |
Seam Type | Serged |
Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126 |