Learn More
DuPont™ Tyvek™ IsoClean™ Gown
188.49 EUR valid until 2024-12-31
Use promo code "22918" to get your promotional price.
191.00 EUR valid until 2024-05-31
Use promo code "21724" to get your promotional price.
Use promo code "22918" to get your promotional price.
191.00 EUR valid until 2024-05-31
Use promo code "21724" to get your promotional price.
Click to view available options
Size:
Medium
2X-Large
Description
- Not clean-processed and not sterilized
- Bound ties
Specifications
Size | 2X-Large |
Color | White |
Cuff Style | Knit |
Model | IC702S00 |
Seam Type | Serged |