Learn More
Schneider Gerd™ Tweezer
Use these forceps for a variety of applications. Schneider Gerd™ Stainless Steel Tweezers are ergonomically designed for optimal grip and precision.
7.60 EUR - 17.85 EUR
Product Code | Brand | Length (Metric) | Design | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Design | Price | Quantity & Availability | ||||
10051214
|
Schneider Gerd™
111 |
115 mm | Blunt |
17.85 EUR
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10284624
|
Schneider Gerd™
125 |
200 mm | Bent |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Stainless steel tweezers
- Straight tips
- Ideal for dissection and general lab applications
Specifications
Tweezers | |
For Laboratory use |