Learn More
SI Analytics TITRONIC 300 Module 1 Basic Unit
Robust, reliable, ready-to-use, TITRONIC 300, Module 1 basic unit that is ideal for use in many laboratory and scientific and research environments.
Brand: SI Analytics 285225810
1803.15 EUR valid until 2024-05-31
Use promo code "21700" to get your promotional price.
Additional Details : Weight : 5.00000kg
Description
- TITRONIC 300 Module 1 TM 300/50 100-240V
- Motor-driven piston burette with 20 or 50 ml dosing unit
- For manual titrations and highly precise dosing tasks
- The TITRONIC ™ 300 can completely controlled from a PC via the USB- or RS-232-interface
- Up to 16 burettes can be connected to one another via the RS232 interface according to the daisy chain principle
Specifications
TITRONIC 300 Module 1 Basic Unit Titrator | |
100-240V |