Learn More
Thermo Scientific™ Wash Buffer for GeneJET Gel Extraction Kit
Product Code. 10509900 Shop All Thermo Scientific Products
Click to view available options
Quantity:
45 mL
Unit Size:
45mL
Product Code. 10509900
Brand: Thermo Scientific™ R1341
Thermo Scientific GeneJET Wash Buffer is a component of the GeneJET Gel Extraction Kit and may be purchased separately. Provided in 45 mL aliquots.
Thermo Scientific GeneJET Wash Buffer is a component of the GeneJET Gel Extraction Kit (Cat No. K0691, K0692) and may be purchased separately. Provided in 45 mL aliquots.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Wash Buffer
For Use With (Equipment) GeneJET Gel Extraction Kit
Product Line GeneJET
Quantity 45 mL
Solution Type Buffer