Learn More
Thermo Scientific™ Cap-It Tool (for domed cap strips)
Make sealing PCR and qPCR plates or tube strips easy
Brand:  Thermo Scientific™ AB0536
41.96 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Additional Details : Weight : 0.04500kg
Product Code. 10203123
  • 44.17 EUR / 1 unit
Estimated Shipment: 03-06-2024
Description
Thermo Scientific Sealing Accessories feature the Cap-IT Tool, which helps you add domed caps to tube strips and plates and helps seal multiple tubes at the same time. The Adhesive Seal Applicator helps you effectively seal PCR and qPCR plates and storage plates.
Highlights
Cap-It Tool
  • Designed to assist in applying domed caps to tube strips and plates
  • It minimizes the pressure required to seal the caps effectively
  • Enables multiple tubes to be sealed simultaneously
Adhesive Seal Applicator
  • Helps ensure effective and consistent sealing of PCR/qPCR plates or storage plates with any adhesive seal
Hazardous:No
Storage Condition: Ambient
Specifications
Specifications
8-Well strips, 96-Well Plates, 96-Well Plate, Fast (0.1 ml), 48-Well Plates, 24-Well Plates