Learn More
Socorex™ Syringe Classic 173 0.3mL to 2mL glass / metal Socorex
Brand:  Socorex™ 173.0502
Additional Details : Weight : 0.10000kg
Product Code. 10223112
  • 133.00 EUR / Each
Estimated Shipment: 03-07-2024