Learn More
Lenz Laborglasintrumente Stirrers with Lateral Blade and GL Thread Connection
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06307035
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11973931
  • 122.96 EUR / Each
Estimated Shipment: 07-06-2024
Description
  • Made of precision calibrated glass tube
  • Length of the ground and polished bearing surface: 160 mm
  • Blade and security
  • Screw: PTFE
  • Shaft made of DURAN™ tubing
Specifications
Specifications
For Joint NS 29/32
390 mm
Duran Glass