Learn More
RETSCH Stainless-Steel Test Sieve, 200 Dia. x 25mmH, Pore sizes in micrometers, ISO 3310/1
Use for separation, fractioning, or particle size determination. These stainless-steel test sieves have an internal height of 25mm and diameter of 200mm. Available in a wide range of micrometer pore sizes.
109.00 EUR - 865.00 EUR
Specifications
Diameter (Metric) Outer | 200 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1 Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10564872
|
RETSCH
60.122.000106 |
106μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10761233
|
RETSCH
60.122.000112 |
112μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10186211
|
RETSCH
60.122.000140 |
140μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10739512
|
RETSCH
60.122.000160 |
160μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10596212
|
RETSCH
60.122.000200 |
200μm |
109.00 EUR
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
10146211
|
RETSCH
60.122.000020 |
20μm |
865.00 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
10196451
|
RETSCH
60.122.000212 |
212μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10014921
|
RETSCH
60.122.000224 |
224μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10330182
|
RETSCH
60.122.000025 |
25μm |
452.00 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
10699911
|
RETSCH
60.122.000280 |
280μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10298921
|
RETSCH
60.122.000300 |
300μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10630102
|
RETSCH
60.122.000315 |
315μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10238491
|
RETSCH
60.122.000032 |
32μm |
234.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10496231
|
RETSCH
60.122.000355 |
355μm |
123.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10574672
|
RETSCH
60.122.000036 |
36μm |
262.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10330862
|
RETSCH
60.122.000400 |
400μm |
109.00 EUR
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
10701233
|
RETSCH
60.122.000038 |
38μm |
262.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10460022
|
RETSCH
60.122.000040 |
40μm |
181.00 EUR
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
10125981
|
RETSCH
60.122.000425 |
425μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10135981
|
RETSCH
60.122.000450 |
450μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10349511
|
RETSCH
60.122.000045 |
45μm |
181.00 EUR
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
10106311
|
RETSCH
60.122.000050 |
50μm |
159.00 EUR
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
10288921
|
RETSCH
60.122.000053 |
53μm |
159.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10258781
|
RETSCH
60.122.000560 |
560μm |
126.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10034801
|
RETSCH
60.122.000056 |
56μm |
142.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10690102
|
RETSCH
60.122.000600 |
600μm |
127.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15630369
|
RETSCH
60.122.000630 |
630μm |
153.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15690259
|
RETSCH
60.122.000630 |
630μm |
169.00 EUR
Each |
Estimated Shipment: 03-09-2024
Log in to see stock. |
|||||
10447581
|
RETSCH
60.122.000710 |
710μm |
126.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10054801
|
RETSCH
60.122.000071 |
71μm |
123.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10711623
|
RETSCH
60.122.000075 |
75μm |
123.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10690082
|
RETSCH
60.122.000080 |
80μm |
123.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10259021
|
RETSCH
60.122.000850 |
850μm |
126.00 EUR
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
10065061
|
RETSCH
60.122.000900 |
900μm |
111.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10600092
|
RETSCH
60.122.000090 |
90μm |
123.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10461973
|
RETSCH
60.122.000180 |
180μm |
123.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Beschreibung
- Stainless-steel frame with wire gauze
- Diameter: 200mm
- Height: 25mm
- ISO 3310/1
Spezifikation
200 mm | |
Stainless Steel | |
1 Each | |
Shaker AS 200 |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
25mm |