Learn More
Haldenwanger™ Squat Form Porcelain Crucibles Without Lid
Use these dishes for a variety of applications. Haldenwanger™ Squat Form Porcelain Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
7.60 EUR - 118.72 EUR
Specifications
Material | Porcelain |
---|---|
Finish | Glazed |
Product Type Specs | Squat Form Porcelain Crucibles |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12306487
|
Haldenwanger™
79/00 |
11 mL |
55.65 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
12316487
|
Haldenwanger™
79/0 |
15 mL |
68.25 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
12326487
|
Haldenwanger™
79/1 |
25 mL |
82.20 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
12336487
|
Haldenwanger™
79/2 |
40 mL |
104.36 EUR
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
10424621
|
Haldenwanger™
L 79/2 |
40 mL |
10.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
12396477
|
Haldenwanger™
79/000A |
6 mL |
54.43 EUR
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12356487
|
Haldenwanger™
79/3 |
85 mL |
79.39 EUR
Pack of 5 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
12346487
|
Haldenwanger™
79/2A |
50 mL |
118.72 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
10665021
|
Haldenwanger™
L 79/2A |
50 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10424431
|
Haldenwanger™
L 79/1 |
25 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Available in a variety of sizes
- Sizes up to 175mL glazed except rim
- 1,000mL and larger are glazed except outside base
- Complies with DIN VDE 0335, type C 100
Specifications
Porcelain | |
Glazed | |
Squat Form Porcelain Crucibles |