missing translation for 'onlineSavingsMsg'
Learn More

SOX17 Monoclonal Antibody (OTI3B10), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA524885

Additional Details : Weight : 0.01000kg

Product Code. 15934494

  • 518.00 EUR / 100µL
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SOX17 (transcription factor SOX-17) belongs to the SRY-related HMB box family of transcription factors. It acts as a transcription regulator that binds target promoter DNA and bends the DNA. It binds to the sequences 5'-AACAAT-'3 or 5'-AACAAAG-3'. SOX17 modulates transcriptional regulation via WNT3A and inhibits Wnt signaling. It promotes degradation of activated CTNNB1. SOX17 plays an essential role in the regulation of embryonic development and is required for normal looping of the embryonic hear tube and for normal development of the definitive gut endoderm. Mutations in the gene can result in vesicoureteral reflux.
Specifications

Specifications

SOX17
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H6I2
SOX17
Human recombinant protein fragment corresponding to amino acids 177-414 of SOX17 produced in E.coli
100μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Western Blot
OTI3B10
Unconjugated
SOX17
HMG transcription factor SOX17; HMG-box transcription factor Sox17; Sox17; Sox-17; SRY (sex determining region Y)-box 17; SRY box 17; SRY-box 17; SRY-box containing gene 17; SRY-box transcription factor 17; SRY-related HMG-box transcription factor SOX17; transcription factor SOX-17; transcription factor SOX-17; LOW QUALITY PROTEIN: transcription factor SOX-17; VUR3
Mouse
Affinity Chromatography
RUO
64321
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers