Learn More
Saint Gobain Life Sciences Silicon Piston Ring
Use this piston ring for a variety of applications. Silicon Piston Ring is available in a full complement of solid and split joint configurations.
23.45 EUR - 260.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15869272
|
Saint Gobain Life Sciences
161040 |
50 to 500 mL Flask |
23.45 EUR
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
15859312
|
Saint Gobain Life Sciences
161042 |
500 to 2000 mL Flask |
29.90 EUR
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
11704608
|
Saint Gobain Life Sciences
161044 |
2 to 6 L Flask |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Sturdy rubber Silicone piston rings for flasks
- For general lab applications
Specifications
Piston Ring | |
Silicon |