Learn More
Epredia™ EZ Megafunnel™ with Caps and Slides
Prepare larger volume samples conveniently with the Epreida™ EZ Megafunnel™ with Caps and Slides.
Brand:  Epredia™ A78710001
Additional Details : Weight : 0.70400kg
Product Code. 12026679
  • 378.00 EUR / Pack of 25
Estimated Shipment: 03-06-2024
Description
  • Minimizes the need to pre-centrifuge
  • Increases cell yield with cell deposition area of 22 x 14.75 mm (325 mm2)
  • Prepares both air-dried and fixed preparations, providing additional valuable diagnostic information
  • Includes Epredia™ EZ Megafunnel™ Cytoslides™
Specifications
Specifications
25/cs.
Megafunnel