Learn More
WTW™ SenTix™ 81 pH Electrode
Use these pH electrodes for portable measurement and routine measurement in the lab. WTW™ SenTix™ 81 pH Electrodes are ideal for applications where reliable measured values are needed under adverse conditions.
Brand: WTW™ 103642
310.27 EUR valid until 2024-05-31
Use promo code "21700" to get your promotional price.
Additional Details : Weight : 0.19260kg
Description
- Precision electrode with temperature sensor and DIN cable
Specifications
Fixed Cable | |
Glass Shaft, Platinum Diaphragm | |
0°C to 100°C | |
Vario pH meter | |
SenTix™ 81 | |
Immerse the combination electrode in reference electrolyte (KCl 3mol/L, Ag+ free), pH combination electrodes must not be stored dry or in deionized water | |
1 m |
Platinum | |
0 to 14 | |
SenTix™ pH Electrode | |
DIN, banana electrode plug, 1m cable | |
30kilohms Built-in NTC | |
0°C to 100°C | |
120 mm |