Learn More
OHAUS™ Sand Bath
Click to view available options
Description:
Sand Bath, 1 Block Unit
Sand Bath, 2 Block Unit
Sand Bath, 4 Block Unit
Sand Bath, 6 Block Unit
Description
- Stainless steel sand baths are available for irregular samples when there is no suitable block available
- Low temperature covers help to reduce the air flow for additional temperature stability
Specifications
Specifications
Description | Sand Bath, 6 Block Unit |
Holds | 6 Blocks |
Material | Stainless Steel |
Type | Accessory, Sand Bath |
Size | 6 Block |