missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Protein LoBind™ Tubes
Protein LoBind™ Tubes ensure optimized sample recovery for improved assay results.
Brand: Eppendorf™ 0030108531
104.00 EUR valid until 2024-12-31
Use promo code "23292" to get your promotional price.
Additional Details : Weight : 0.25000kg
Description
- Eppendorf LoBind material ensures optimized sample recovery for improved assay results
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
- Available in tube, microplate, and deepwell plate formats for easy-up scaling
- Precise lid sealing to minimize evaporation
Specifications
Colorless | |
100/Bag, 2 Bags | |
PCR Clean | |
Snap Cap |
25 mL | |
200 Tubes | |
Conical | |
Tube |