Learn More
3M™ Over-the-Glass Visitor Safety Glasses
1.95 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
1.95 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "21742" to get your promotional price.
1.95 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Click to view available options
Clear
Description
Classic style visitor spectacles
- Lightweight lenses
- Moulded sideshields and browguards
- Can be worn on their own or over current style prescription spectacles.
- Integrated side-shields for maximum protection
- Lens coatings provide scratch resistance
Specifications
Lens Tint | Clear |
Product Type | Protective eyewear |
For Use With (Application) | Protection Against Low Energy Impact, UV Protection |
UV Protection | Yes |