Learn More
Microspec™ Plastic Inoculation Loops
Available as aseptic manufactured or ethylene oxide sterilized.
26.90 EUR - 28.90 EUR
Specifications
Disposable | Yes |
---|---|
Color | Blue |
Length (Metric) Handle | 200 mm |
Packaging Type | Microseal Pouch |
Autoclavable | No |
Product Code | Brand | Volume | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume | Sterility | Price | Quantity & Availability | ||||
16664822
|
Microspec™
C5G |
1 μL | Sterile |
26.90 EUR
Pack of 250 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
16644822
|
Microspec™
B5G |
5 μL | Sterile |
26.90 EUR
Pack of 250 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
16674782
|
Microspec™
A5G |
10 μL | Sterile |
26.90 EUR
Pack of 250 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
16654822
|
Microspec™
C5 |
1 μL | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16664782
|
Microspec™
A5 |
10 μL | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16634822
|
Microspec™
B5 |
5 μL | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Includes 250 Plastic Inoculation Loops
- Aseptic Manufactured or Ethylene Oxide Sterilized
- 200 mm length
Specifications
Yes | |
200 mm | |
No |
Blue | |
Microseal Pouch |