missing translation for 'onlineSavingsMsg'
Learn More

Perilipin 1 Recombinant Rabbit Monoclonal Antibody (JM10-96), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen MA532597

Additional Details : Weight : 0.01000kg

Product Code. 16376004

  • 471.00 EUR / 100µL
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Adipose tissue is an energy reserve in animals and is strictly regulated in nondomestic species. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), resistin, and perilipin. Perilipin is an intracellular neutral lipid droplet protein that is hormonally regulated. This protein is localized exclusively to the surface of lipid droplets. In response to lypotic stimuli, perilipin is phosphorylated by protein kinase A. Once activated, perilipin has inhibitory affects upon hormone-sensitive lipase (HSL), a protein that mediates the hydrolysis of triacylglycerol, the major form of stored energy in the body. Perilipin expression is limited to adipocytes and steroidogenic cells. There are currently two known isoforms, Perilipin A and B. Both of these proteins are encoded by a single-copy gene and are the result of differential splicing events.
Specifications

Specifications

Perilipin 1
Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide, pH 7.4
O60240, Q8CGN5
Plin1
Synthetic peptide within Human Perilipin A aa 470-510
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunocytochemistry, Western Blot, Western Blot
JM10-96
Unconjugated
Plin1
6030432J05Rik, adipocyte lipid droplet binding protein, FPLD4, Lipid droplet-associated protein, PERI, PERIA, perilipin, perilipin 1, perilipin A, perilipin B, perilipin-1, PLIN, PLIN1
Rabbit
Protein A
RUO
103968, 5346
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers